Product Details

SNP ID
rs138966068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36169750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTACCTGAGCCCGGTCACAGTGG[A/G]CCAGCTTCTGGACTACTTCTATAGC
Phenotype
MIM: 603960 MIM: 607141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCIN PubMed Links

Gene Details

Gene
CCIN
Gene Name
calicin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005893.2 359 Missense Mutation GAC,GGC D83G NP_005884.2
Gene
GLIPR2
Gene Name
GLI pathogenesis related 2
There are no transcripts associated with this gene.

View Full Product Details