Product Details

SNP ID
rs138992924
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91409910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTACCACACCATCATCAGTTTC[C/T]GACGTTCTCGGGGAGTTGCTGGAGG
Phenotype
MIM: 605327
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NFIL3 PubMed Links

Gene Details

Gene
NFIL3
Gene Name
nuclear factor, interleukin 3 regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289999.1 1358 Silent Mutation NP_001276928.1
NM_001290000.1 1358 Silent Mutation NP_001276929.1
NM_005384.2 1358 Silent Mutation NP_005375.2
XM_017014743.1 1358 Silent Mutation XP_016870232.1
XM_017014744.1 1358 Silent Mutation XP_016870233.1

View Full Product Details