Product Details

SNP ID
rs142355207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136208880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGTCAACCCCGAGGAAGGGTG[C/T]GGCCCCGCCCACCTCCTTGTGGGCC
Phenotype
MIM: 600577 MIM: 612860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHX3 PubMed Links

Gene Details

Gene
LHX3
Gene Name
LIM homeobox 3
There are no transcripts associated with this gene.

Gene
QSOX2
Gene Name
quiescin sulfhydryl oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181701.3 1983 Missense Mutation ACA,GCA T649A NP_859052.3

View Full Product Details