Product Details

SNP ID
rs144423836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128345436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCCTATGGCTGGGCATGGCCA[A/G]GCTCGGTGTGGAGGCAGCCCTCATC
Phenotype
MIM: 604194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC27A4 PubMed Links

Gene Details

Gene
SLC27A4
Gene Name
solute carrier family 27 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005094.3 601 Missense Mutation AAG,AGG K148R NP_005085.2
XM_017014222.1 601 Missense Mutation AAG,AGG K148R XP_016869711.1

View Full Product Details