Product Details

SNP ID
rs144538828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122519288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCTGCTCATCATCCTGCTCATC[C/T]GGCTGGACTCTCACCTTCACACCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1J2 PubMed Links

Gene Details

Gene
OR1J2
Gene Name
olfactory receptor family 1 subfamily J member 2
There are no transcripts associated with this gene.

Gene
OR1J4
Gene Name
olfactory receptor family 1 subfamily J member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004452.1 148 Missense Mutation CGG,TGG R50W NP_001004452.1
Gene
OR1N1
Gene Name
olfactory receptor family 1 subfamily N member 1
There are no transcripts associated with this gene.

View Full Product Details