Product Details

SNP ID
rs147007838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:88542616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATGGTTTCTTATTGGTTTCCA[A/G]TTCCATTACCATTGGTCTGATTAAG
Phenotype
MIM: 615299
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NXNL2 PubMed Links

Gene Details

Gene
NXNL2
Gene Name
nucleoredoxin-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161625.1 Intron NP_001155097.1
NM_145283.2 Intron NP_660326.2
XM_005251727.3 Intron XP_005251784.1
XM_011518275.2 Intron XP_011516577.1
XM_011518276.2 Intron XP_011516578.1

View Full Product Details