Product Details

SNP ID
rs147457502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133098827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCCTGCAGGATACCCCTACC[A/G]CTTGAGAGCCCATACAAGGACTGTC
Phenotype
MIM: 601619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RALGDS PubMed Links

Gene Details

Gene
RALGDS
Gene Name
ral guanine nucleotide dissociation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042368.2 Intron NP_001035827.1
NM_001271774.1 Intron NP_001258703.1
NM_001271775.1 Intron NP_001258704.1
NM_001271776.1 Intron NP_001258705.1
NM_006266.3 Intron NP_006257.1

View Full Product Details