Product Details

SNP ID
rs148808709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35075531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCAGGCCTGGCCCTGAAGCAGG[A/T]GGGTGGCAGAGACCCAGAGTGGGCA
Phenotype
MIM: 602956 MIM: 614730 MIM: 601023
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FANCG PubMed Links

Gene Details

Gene
FANCG
Gene Name
Fanconi anemia complementation group G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004629.1 1859 Missense Mutation CAC,CTC H456L NP_004620.1
Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
There are no transcripts associated with this gene.

Gene
VCP
Gene Name
valosin containing protein
There are no transcripts associated with this gene.

View Full Product Details