Product Details

SNP ID
rs149000990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34253339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGGCTTTAAAGTAATTGGCAC[C/T]TGGAGGCTACTTTGTCTTCCCTGTC
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 3872 UTR 3 NP_919289.2
XM_005251451.2 3872 UTR 3 XP_005251508.1
XM_006716764.2 3872 UTR 3 XP_006716827.1
XM_011517861.1 3872 UTR 3 XP_011516163.1
XM_011517863.2 3872 UTR 3 XP_011516165.1
XM_017014697.1 3872 UTR 3 XP_016870186.1
XM_017014698.1 3872 UTR 3 XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171201.1 3872 Intron NP_001164672.1
NM_001171202.1 3872 Intron NP_001164673.1
NM_001171203.2 3872 Intron NP_001164674.1
NM_001171204.2 3872 Intron NP_001164675.1
NM_016525.4 3872 Intron NP_057609.2
XM_006716779.3 3872 Intron XP_006716842.1
XM_011517898.2 3872 Intron XP_011516200.1
XM_011517899.1 3872 Intron XP_011516201.1
XM_011517900.1 3872 Intron XP_011516202.1
XM_017014801.1 3872 Intron XP_016870290.1
XM_017014802.1 3872 Intron XP_016870291.1

View Full Product Details