Product Details

SNP ID
rs149697542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21077800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTAGGAATCCAAGCAAGTTGT[A/G]GCTCATGGAAAGAGCTGTAGTGGAG
Phenotype
MIM: 147640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNB1 PubMed Links

Gene Details

Gene
IFNB1
Gene Name
interferon beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002176.3 164 Missense Mutation CAC,TAC H24Y NP_002167.1

View Full Product Details