Product Details
- SNP ID
-
rs140304472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:20010875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCGATAAGGTTTTTGTTACAGTC[A/G]TCCAGGCTGAGATTTTCACTGGATC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP7D2
PubMed Links
Gene Details
- Gene
- MAP7D2
- Gene Name
- MAP7 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001168465.1 |
2139 |
Silent Mutation |
GAC,GAT |
D750D |
NP_001161937.1 |
NM_001168466.1 |
2139 |
Silent Mutation |
GAC,GAT |
D664D |
NP_001161938.1 |
NM_001168467.1 |
2139 |
Silent Mutation |
GAC,GAT |
D657D |
NP_001161939.1 |
NM_152780.3 |
2139 |
Silent Mutation |
GAC,GAT |
D709D |
NP_689993.2 |
XM_005274478.2 |
2139 |
Silent Mutation |
GAC,GAT |
D734D |
XP_005274535.1 |
XM_005274479.2 |
2139 |
Silent Mutation |
GAC,GAT |
D723D |
XP_005274536.1 |
XM_005274480.3 |
2139 |
Silent Mutation |
GAC,GAT |
D701D |
XP_005274537.1 |
XM_006724481.2 |
2139 |
Silent Mutation |
GAC,GAT |
D783D |
XP_006724544.1 |
XM_006724482.2 |
2139 |
Silent Mutation |
GAC,GAT |
D742D |
XP_006724545.1 |
XM_011545483.1 |
2139 |
Silent Mutation |
GAC,GAT |
D772D |
XP_011543785.1 |
XM_011545484.1 |
2139 |
Silent Mutation |
GAC,GAT |
D739D |
XP_011543786.1 |
XM_011545485.1 |
2139 |
Silent Mutation |
GAC,GAT |
D728D |
XP_011543787.1 |
XM_011545486.2 |
2139 |
Intron |
|
|
XP_011543788.1 |
XM_017029393.1 |
2139 |
Silent Mutation |
GAC,GAT |
D731D |
XP_016884882.1 |
XM_017029394.1 |
2139 |
Silent Mutation |
GAC,GAT |
D698D |
XP_016884883.1 |
XM_017029395.1 |
2139 |
Silent Mutation |
GAC,GAT |
D690D |
XP_016884884.1 |
XM_017029396.1 |
2139 |
Silent Mutation |
GAC,GAT |
D687D |
XP_016884885.1 |
XM_017029397.1 |
2139 |
Silent Mutation |
GAC,GAT |
D679D |
XP_016884886.1 |
XM_017029398.1 |
2139 |
Intron |
|
|
XP_016884887.1 |
- Gene
- MIR23C
- Gene Name
- microRNA 23c
There are no transcripts associated with this gene.
View Full Product Details