Product Details

SNP ID
rs141926826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151404934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATAGGCGCCCTTGGGATGTCCA[A/G]TAGGGACAGCTATTTTTACGCTGCT
Phenotype
MIM: 300913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VMA21 PubMed Links

Gene Details

Gene
VMA21
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017980.3 632 Missense Mutation AAT,AGT N61S NP_001017980.1
XM_011531125.2 632 Missense Mutation AAT,AGT N116S XP_011529427.1

View Full Product Details