Product Details

SNP ID
rs143293969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120370729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCTGCAGGCCTGATCTTACTC[A/G]TTTACTTCTTCTTCTATGCCTCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1B4 PubMed Links

Gene Details

Gene
ATP1B4
Gene Name
ATPase Na+/K+ transporting family member beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142447.2 427 Missense Mutation ATT,GTT I115V NP_001135919.1
NM_012069.4 427 Missense Mutation ATT,GTT I111V NP_036201.1
XM_017029381.1 427 Missense Mutation ATT,GTT I115V XP_016884870.1

View Full Product Details