Product Details
- SNP ID
-
rs144099565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:25015600 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCGGAGCAGGCAAGCTCTGCGC[G/T]GCTCCCAGCAACCGCATTTTGCACG
- Phenotype
-
MIM: 300382
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARX
PubMed Links
Gene Details
- Gene
- ARX
- Gene Name
- aristaless related homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139058.2 |
349 |
Silent Mutation |
GCA,GCC |
A46A |
NP_620689.1 |
View Full Product Details