Product Details

SNP ID
rs144277194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100987344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACTGGTTCTGCGGGCCCCAGG[A/G]GTCCCAGGCTCCAGGTCTGCAGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL13A PubMed Links

Gene Details

Gene
ARL13A
Gene Name
ADP ribosylation factor like GTPase 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162491.1 Intron NP_001155963.1
XM_011530950.2 Intron XP_011529252.1
XM_011530951.2 Intron XP_011529253.1
Gene
TRMT2B
Gene Name
tRNA methyltransferase 2 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167970.1 Intron NP_001161442.1
NM_001167971.1 Intron NP_001161443.1
NM_001167972.1 Intron NP_001161444.1
NM_024917.5 Intron NP_079193.2
XM_005262195.2 Intron XP_005262252.1
XM_005262196.2 Intron XP_005262253.1
XM_005262201.2 Intron XP_005262258.1
XM_006724705.2 Intron XP_006724768.1
XM_011531042.2 Intron XP_011529344.1
XM_011531044.2 Intron XP_011529346.1
XM_017029858.1 Intron XP_016885347.1
XM_017029859.1 Intron XP_016885348.1
XM_017029860.1 Intron XP_016885349.1
XM_017029861.1 Intron XP_016885350.1
XM_017029862.1 Intron XP_016885351.1
XM_017029863.1 Intron XP_016885352.1
XM_017029864.1 Intron XP_016885353.1
XM_017029865.1 Intron XP_016885354.1
XM_017029866.1 Intron XP_016885355.1
XM_017029867.1 Intron XP_016885356.1
XM_017029868.1 Intron XP_016885357.1
XM_017029869.1 Intron XP_016885358.1
XM_017029870.1 Intron XP_016885359.1
XM_017029871.1 Intron XP_016885360.1
XM_017029872.1 Intron XP_016885361.1
XM_017029873.1 Intron XP_016885362.1
XM_017029874.1 Intron XP_016885363.1
XM_017029875.1 Intron XP_016885364.1

View Full Product Details