Product Details
- SNP ID
-
hCV175447154
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101657225 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGATTCGGCCTTAGGCCCAACCC[A/C]GGCTCCATCTGCCTCTTGGGCCTGA
- Phenotype
-
MIM: 300363
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARMCX2
PubMed Links
Gene Details
- Gene
- ARMCX2
- Gene Name
- armadillo repeat containing, X-linked 2
There are no transcripts associated with this gene.
View Full Product Details