Product Details

SNP ID
rs145922470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43767791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTCTTCCTCACTGGACCCTTCA[C/T]TTCCTCAGGGAGGCTTTCTTTCCTG
Phenotype
MIM: 309860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAOB PubMed Links

Gene Details

Gene
MAOB
Gene Name
monoamine oxidase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000898.4 Intron NP_000889.3
XM_005272608.3 Intron XP_005272665.1
XM_017029523.1 Intron XP_016885012.1
XM_017029524.1 Intron XP_016885013.1

View Full Product Details