Product Details
- SNP ID
-
rs146364929
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135287378 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTGTGTAGATTTGTGTTATGA[C/G]TAAAGCTTTTCCCACACCATGAGCA
- Phenotype
-
MIM: 314997
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF75D
PubMed Links
Gene Details
- Gene
- ZNF75D
- Gene Name
- zinc finger protein 75D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001185063.2 |
3131 |
Intron |
|
|
NP_001171992.1 |
NM_007131.4 |
3131 |
Missense Mutation |
ACT,AGT |
T431S |
NP_009062.2 |
XM_005262469.1 |
3131 |
Missense Mutation |
ACT,AGT |
T431S |
XP_005262526.1 |
XM_005262471.3 |
3131 |
Missense Mutation |
ACT,AGT |
T224S |
XP_005262528.1 |
XM_011531391.1 |
3131 |
Missense Mutation |
ACT,AGT |
T431S |
XP_011529693.1 |
XM_011531395.2 |
3131 |
Missense Mutation |
ACT,AGT |
T281S |
XP_011529697.1 |
XM_011531396.2 |
3131 |
Missense Mutation |
ACT,AGT |
T263S |
XP_011529698.1 |
XM_011531397.2 |
3131 |
Missense Mutation |
ACT,AGT |
T260S |
XP_011529699.1 |
XM_017029818.1 |
3131 |
Missense Mutation |
ACT,AGT |
T285S |
XP_016885307.1 |
XM_017029819.1 |
3131 |
Missense Mutation |
ACT,AGT |
T297S |
XP_016885308.1 |
XM_017029820.1 |
3131 |
Missense Mutation |
ACT,AGT |
T260S |
XP_016885309.1 |
View Full Product Details