Product Details

SNP ID
rs147795953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135292465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGCCTCCTTTCCCAGCTCATG[G/T]GCTGTGACCTAGAAATAATCCCCAT
Phenotype
MIM: 314997
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF75D PubMed Links

Gene Details

Gene
ZNF75D
Gene Name
zinc finger protein 75D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185063.2 2351 Intron NP_001171992.1
NM_007131.4 2351 Silent Mutation GCA,GCC A140A NP_009062.2
XM_005262469.1 2351 Silent Mutation GCA,GCC A140A XP_005262526.1
XM_005262471.3 2351 UTR 5 XP_005262528.1
XM_011531391.1 2351 Silent Mutation GCA,GCC A140A XP_011529693.1
XM_011531395.2 2351 Missense Mutation CAC,CCC H21P XP_011529697.1
XM_011531396.2 2351 UTR 5 XP_011529698.1
XM_011531397.2 2351 UTR 5 XP_011529699.1
XM_017029818.1 2351 Missense Mutation CAC,CCC H25P XP_016885307.1
XM_017029819.1 2351 Missense Mutation CAC,CCC H37P XP_016885308.1
XM_017029820.1 2351 UTR 5 XP_016885309.1

View Full Product Details