Product Details
- SNP ID
-
hCV175626615
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:154695922 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGAAAAATATAAGATTACCATTT[C/T]GATGTAGCTTTCTTCGTCTTCTCTG
- Phenotype
-
MIM: 300482
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAB3
PubMed Links
Gene Details
- Gene
- GAB3
- Gene Name
- GRB2 associated binding protein 3
There are no transcripts associated with this gene.
View Full Product Details