Product Details

SNP ID
rs150635004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2243047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAAAAGCAACCAGCCGAGAAG[C/T]TTCTTCGCCAGACGGGTGGCCCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PubMed Links

Gene Details

View Full Product Details