Product Details

SNP ID
rs11547706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44799799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATCCTAAAGCATACGGGTCCTG[A/G]CATCTTGTCCATGGCAAATGCTGGA
Phenotype
MIM: 123840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIA PubMed Links

Gene Details

Gene
PPIA
Gene Name
peptidylprolyl isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300981.1 376 Intron NP_001287910.1
NM_021130.4 376 Missense Mutation GAC,GGC D96G NP_066953.1

View Full Product Details