Product Details

SNP ID
rs11548913
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:174999671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGACTCTCTAGTCAACTTCCG[A/T]CTTGGACTCCGAAGATCGGTACGTT
Phenotype
MIM: 606186 MIM: 609238
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CACYBP PubMed Links

Gene Details

Gene
CACYBP
Gene Name
calcyclin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007214.1 237 UTR 5 NP_001007215.1
NM_014412.2 237 Intron NP_055227.1
XM_017001046.1 237 Intron XP_016856535.1
Gene
LOC105371622
Gene Name
uncharacterized LOC105371622
There are no transcripts associated with this gene.

Gene
RABGAP1L
Gene Name
RAB GTPase activating protein 1 like
There are no transcripts associated with this gene.

View Full Product Details