Product Details

SNP ID
rs11547705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44799818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGCATCTTGTCCATGGCAAA[C/T]GCTGGACCCAACACAAATGGTTCCC
Phenotype
MIM: 123840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIA PubMed Links

Gene Details

Gene
PPIA
Gene Name
peptidylprolyl isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300981.1 395 Intron NP_001287910.1
NM_021130.4 395 Silent Mutation AAC,AAT N102N NP_066953.1

View Full Product Details