Product Details

SNP ID
hCV176048665
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53228218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCCCATCCTACAGATGAGGAAA[A/C]TGAAGCTCAGGGAAGTGTAAACTTA
Phenotype
MIM: 300522 MIM: 314690
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
IQSEC2 PubMed Links

Gene Details

Gene
IQSEC2
Gene Name
IQ motif and Sec7 domain 2
There are no transcripts associated with this gene.

Gene
KDM5C
Gene Name
lysine demethylase 5C
There are no transcripts associated with this gene.

View Full Product Details