Product Details

SNP ID
rs12927
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:217284160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGACTTGGTCCATTGATTTGTT[C/T]ATTCACTAATTCAACAACATTTACT
Phenotype
MIM: 608262
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DIRC3 PubMed Links

Gene Details

Gene
DIRC3
Gene Name
disrupted in renal carcinoma 3
There are no transcripts associated with this gene.

Gene
DIRC3-AS1
Gene Name
DIRC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details