Product Details

SNP ID
rs181896105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101035524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGTGGTGACCCCAGGGATCAC[C/T]GAGGACCAGCTGTGGAGGGCCAAGT
Phenotype
MIM: 612971 MIM: 615571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDZD7 PubMed Links

Gene Details

Gene
PDZD7
Gene Name
PDZ domain containing 7
There are no transcripts associated with this gene.

Gene
SFXN3
Gene Name
sideroflexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030971.3 657 Silent Mutation ACC,ACT T67T NP_112233.2

View Full Product Details