Product Details

SNP ID
rs182178642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56319662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGTCATCAATGAAGGGGATAC[A/C]CTCATTGGGTCCCTGGCCCCGAACA
Phenotype
MIM: 605861
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3438 Missense Mutation GGT,TGT G1021C NP_001120932.1
NM_001166279.1 3438 Missense Mutation GGT,TGT G1020C NP_001159751.1
NM_014871.4 3438 Missense Mutation GGT,TGT G1017C NP_055686.3

View Full Product Details