Product Details
- SNP ID
-
rs182608815
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:42029058 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGGCTGGAGTGGAGGCCCAAGA[A/G]GAGGGGCTGCAGGAACTGCTGCAGA
- Phenotype
-
MIM: 601964
MIM: 604497
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJC7
PubMed Links
Gene Details
- Gene
- DNAJC7
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C7
There are no transcripts associated with this gene.
- Gene
- NKIRAS2
- Gene Name
- NFKB inhibitor interacting Ras like 2
There are no transcripts associated with this gene.
- Gene
- ZNF385C
- Gene Name
- zinc finger protein 385C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242704.1 |
1385 |
Missense Mutation |
CCT,CTT |
P152L |
NP_001229633.1 |
XM_006721751.3 |
1385 |
Missense Mutation |
CCT,CTT |
P99L |
XP_006721814.1 |
XM_011524475.2 |
1385 |
Intron |
|
|
XP_011522777.1 |
XM_011524476.1 |
1385 |
Intron |
|
|
XP_011522778.1 |
XM_011524477.2 |
1385 |
Intron |
|
|
XP_011522779.1 |
XM_011524478.2 |
1385 |
Intron |
|
|
XP_011522780.1 |
XM_011524479.2 |
1385 |
Missense Mutation |
CCT,CTT |
P232L |
XP_011522781.1 |
XM_011524482.2 |
1385 |
Missense Mutation |
CCT,CTT |
P231L |
XP_011522784.1 |
XM_011524483.2 |
1385 |
Missense Mutation |
CCT,CTT |
P232L |
XP_011522785.1 |
XM_017024314.1 |
1385 |
Missense Mutation |
CCT,CTT |
P274L |
XP_016879803.1 |
XM_017024315.1 |
1385 |
Missense Mutation |
CCT,CTT |
P274L |
XP_016879804.1 |
XM_017024316.1 |
1385 |
Missense Mutation |
CCT,CTT |
P273L |
XP_016879805.1 |
XM_017024317.1 |
1385 |
Missense Mutation |
CCT,CTT |
P274L |
XP_016879806.1 |
XM_017024318.1 |
1385 |
Missense Mutation |
CCT,CTT |
P273L |
XP_016879807.1 |
XM_017024319.1 |
1385 |
Missense Mutation |
CCT,CTT |
P231L |
XP_016879808.1 |
XM_017024320.1 |
1385 |
Missense Mutation |
CCT,CTT |
P141L |
XP_016879809.1 |
XM_017024321.1 |
1385 |
UTR 3 |
|
|
XP_016879810.1 |
View Full Product Details