Product Details

SNP ID
rs183157251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105339636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAAACAAAGACAGTCTTACTGA[C/T]CTTTATGTCCAACATGCAATACCAT
Phenotype
MIM: 606237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf49 PubMed Links

Gene Details

Gene
C2orf49
Gene Name
chromosome 2 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286537.1 484 Silent Mutation GAC,GAT D51D NP_001273466.1
NM_024093.2 484 Silent Mutation GAC,GAT D51D NP_076998.1
XM_017004892.1 484 Silent Mutation GAC,GAT D89D XP_016860381.1
Gene
LOC100506473
Gene Name
uncharacterized LOC100506473
There are no transcripts associated with this gene.

Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
There are no transcripts associated with this gene.

View Full Product Details