Product Details

SNP ID
rs183181300
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:53459683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTGCGCAGGGGCCCAAGCAGG[C/G]CTCCGGGGCTGCGGCCGCCGCCCCC
Phenotype
MIM: 614805
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DMRTB1 PubMed Links

Gene Details

Gene
DMRTB1
Gene Name
DMRT like family B with proline rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033067.2 285 Missense Mutation GCC,GGC A77G NP_149056.1

View Full Product Details