Product Details
- SNP ID
-
rs183545746
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85439854 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGATGGTGGCTCTGCCAAGACCAT[A/C]CTGAATAATGGGAAGACCTGCCGAG
- Phenotype
-
MIM: 114750
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CA3
PubMed Links
Gene Details
- Gene
- CA3
- Gene Name
- carbonic anhydrase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005181.3 |
260 |
Silent Mutation |
ATA,ATC |
I59I |
NP_005172.1 |
- Gene
- CA3-AS1
- Gene Name
- CA3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details