Product Details

SNP ID
rs183534412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75435458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACACTTCCACCAGCTCTCCTA[A/G]GCTCCCCAGCTTCTGTGTCAGGTAC
Phenotype
MIM: 609632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD5 PubMed Links

Gene Details

Gene
GDPD5
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030792.6 3139 UTR 3 NP_110419.5
XM_006718697.3 3139 UTR 3 XP_006718760.1
XM_011545276.2 3139 UTR 3 XP_011543578.1
XM_011545277.2 3139 UTR 3 XP_011543579.1
XM_011545278.2 3139 UTR 3 XP_011543580.1
XM_011545279.2 3139 UTR 3 XP_011543581.1
XM_011545280.1 3139 UTR 3 XP_011543582.1
XM_011545281.2 3139 UTR 3 XP_011543583.1
XM_011545285.2 3139 UTR 3 XP_011543587.1
XM_011545286.1 3139 UTR 3 XP_011543588.1
XM_011545287.1 3139 UTR 3 XP_011543589.1
Gene
KLHL35
Gene Name
kelch like family member 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039548.2 3139 Intron NP_001034637.2
XM_011544954.2 3139 Intron XP_011543256.1

View Full Product Details