Product Details

SNP ID
rs113641726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133579094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGAGAGGCGGTTGGGGTTGAG[A/C]GCCTGCAGGCCCCCGAAGCCCCCCG
Phenotype
MIM: 612277
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTSL2 PubMed Links

Gene Details

Gene
ADAMTSL2
Gene Name
ADAMTS like 2
There are no transcripts associated with this gene.

Gene
FAM163B
Gene Name
family with sequence similarity 163 member B
There are no transcripts associated with this gene.

View Full Product Details