Product Details

SNP ID
rs184693277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168666590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACTGGGGCCCACAGCCCCCA[C/T]GACATTCCATGATGGGCACCTTGGA
Phenotype
MIM: 603745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLIT3 PubMed Links

Gene Details

Gene
SLIT3
Gene Name
slit guidance ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271946.1 4856 Missense Mutation NP_001258875.1
NM_003062.3 4856 Missense Mutation NP_003053.1
XM_017009779.1 4856 Missense Mutation XP_016865268.1

View Full Product Details