Product Details

SNP ID
rs184826298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35675802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGTGTCCCCAGCCTGCGCGGGC[A/C]GCTTCCAGTCCCCGGTGGATATCCG
Phenotype
MIM: 603179 MIM: 190990
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CA9 PubMed Links

Gene Details

Gene
CA9
Gene Name
carbonic anhydrase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001216.2 517 Missense Mutation AGC,CGC S159R NP_001207.2
Gene
TPM2
Gene Name
tropomyosin 2 (beta)
There are no transcripts associated with this gene.

View Full Product Details