Product Details

SNP ID
rs185601901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTGGCTGTGGCTGGGGCTGGTG[A/T]AGCTGGGCTGTGCCATGGCGTGCCT
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 608 Intron NP_079113.2
XM_011522046.2 608 Intron XP_011520348.1
XM_011522047.2 608 Intron XP_011520349.1
XM_011522048.1 608 Intron XP_011520350.1
XM_011522049.2 608 Intron XP_011520351.1
XM_011522051.2 608 Intron XP_011520353.1
XM_011522052.2 608 Intron XP_011520354.1
XM_011522053.1 608 Intron XP_011520355.1
XM_011522056.2 608 Intron XP_011520358.2
XM_011522058.2 608 Intron XP_011520360.1
XM_011522059.1 608 Intron XP_011520361.1
XM_011522060.1 608 Intron XP_011520362.1
XM_011522061.1 608 Intron XP_011520363.1
XM_011522062.1 608 Intron XP_011520364.1
XM_011522063.1 608 Intron XP_011520365.1
XM_011522064.1 608 Intron XP_011520366.1
XM_011522069.2 608 Intron XP_011520371.1
XM_011522070.1 608 Intron XP_011520372.1
XM_017022587.1 608 Intron XP_016878076.1
XM_017022588.1 608 Intron XP_016878077.1
XM_017022589.1 608 Intron XP_016878078.1
XM_017022590.1 608 Intron XP_016878079.1
XM_017022591.1 608 Intron XP_016878080.1
XM_017022592.1 608 Intron XP_016878081.1
XM_017022593.1 608 Intron XP_016878082.1
XM_017022594.1 608 Intron XP_016878083.1
XM_017022595.1 608 Intron XP_016878084.1
XM_017022596.1 608 Intron XP_016878085.1
XM_017022597.1 608 Intron XP_016878086.1
XM_017022598.1 608 Intron XP_016878087.1
XM_017022599.1 608 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 608 Nonsense Mutation AAG,TAG K126* NP_001153101.1
NM_003645.3 608 Nonsense Mutation AAG,TAG K126* NP_003636.2

View Full Product Details