Product Details

SNP ID
rs185516764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96051209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCCAGTCTGCCGTTGCCACT[G/T]GGAGTTCTTCCAGGTATCCTGTGAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C14orf132 PubMed Links

Gene Details

Gene
C14orf132
Gene Name
chromosome 14 open reading frame 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252507.2 290 Intron NP_001239436.1
NM_001282463.1 290 Missense Mutation TGG,TTG W14L NP_001269392.1
NM_001282464.1 290 Intron NP_001269393.1
NM_001289139.1 290 Intron NP_001276068.1
XM_017021469.1 290 Missense Mutation TGG,TTG W14L XP_016876958.1

View Full Product Details