Product Details

SNP ID
rs185118426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98498453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTCTATGGATCTATTTTTTTCC[A/T]ATACATTAGACCAAATTTGCTTGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5K2 PubMed Links

Gene Details

Gene
OR5K2
Gene Name
olfactory receptor family 5 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004737.1 773 Missense Mutation CAA,CTA Q258L NP_001004737.1

View Full Product Details