Product Details

SNP ID
rs185923262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112980042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTATCACATGCAAGTATCCAG[A/G]GGCTCTTGAGCAAGGCAGAGGGGAT
Phenotype
MIM: 605542
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL36G PubMed Links

Gene Details

Gene
IL36G
Gene Name
interleukin 36, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278568.1 273 Missense Mutation GAG,GGG E30G NP_001265497.1
NM_019618.3 273 Missense Mutation GAG,GGG E65G NP_062564.1

View Full Product Details