Product Details

SNP ID
rs186289957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25417157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTGATACCAACTATCAAATTTC[A/G]ACACTGTTCTCTGTTAAGAAATCGA
Phenotype
MIM: 610608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GINS1 PubMed Links

Gene Details

Gene
GINS1
Gene Name
GINS complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021067.4 289 Missense Mutation CAA,CGA Q65R NP_066545.3
XM_017028162.1 289 Missense Mutation CAA,CGA Q30R XP_016883651.1

View Full Product Details