Product Details

SNP ID
rs186512421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:104855597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCACTCGGACGCGGCAGGGGC[C/T]GGCGGCGGCGGGGGTGGCGGCGGCG
Phenotype
MIM: 602480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01158 PubMed Links

Gene Details

Gene
LINC01158
Gene Name
long intergenic non-protein coding RNA 1158
There are no transcripts associated with this gene.

Gene
LINC01159
Gene Name
long intergenic non-protein coding RNA 1159
There are no transcripts associated with this gene.

Gene
POU3F3
Gene Name
POU class 3 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006236.2 1530 Silent Mutation GCC,GCT A29A NP_006227.1

View Full Product Details