Product Details

SNP ID
rs186425726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10725808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACGGAGATGAGGCTGGTCTCT[A/G]TGGGACCCGTTGGTGTCACCTGGGA
Phenotype
MIM: 612202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723313 PubMed Links

Gene Details

Gene
LOC102723313
Gene Name
uncharacterized LOC102723313
There are no transcripts associated with this gene.

Gene
SOX7
Gene Name
SRY-box 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031439.3 1238 Missense Mutation ACA,ATA T366I NP_113627.1

View Full Product Details