Product Details

SNP ID
rs186203433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135985540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCGGCGGGCACCCCTCCGCC[A/G]TGGCGTCGGCAGCAGTCCCCGAGCC
Phenotype
MIM: 300231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC9A6 PubMed Links

Gene Details

Gene
SLC9A6
Gene Name
solute carrier family 9 member A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042537.1 114 Missense Mutation CAT,CGT H13R NP_001036002.1
NM_001177651.1 114 Intron NP_001171122.1
NM_006359.2 114 Missense Mutation CAT,CGT H13R NP_006350.1
XM_006724726.3 114 Intron XP_006724789.1
XM_011531243.2 114 Intron XP_011529545.1
XM_017029223.1 114 Intron XP_016884712.1
XM_017029224.1 114 Intron XP_016884713.1
XM_017029225.1 114 Intron XP_016884714.1

View Full Product Details