Product Details

SNP ID
rs186199748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44356222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAATGCTGGTGCCCACGGTGAG[A/G]GCACGAGTCTCGGAGGGCACGGGCA
Phenotype
MIM: 138945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198475.2 449 Silent Mutation GCC,GCT A243A NP_940877.2
XM_017024490.1 449 Silent Mutation GCC,GCT A59A XP_016879979.1
Gene
GRN
Gene Name
granulin
There are no transcripts associated with this gene.

View Full Product Details