Product Details

SNP ID
rs187134186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33034213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGTGAAGGATGTCCAGCTGCC[C/T]GTCATCACACTGAACTTTGTACCTG
Phenotype
MIM: 614110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFB6 PubMed Links

Gene Details

Gene
BPIFB6
Gene Name
BPI fold containing family B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174897.2 225 Silent Mutation CCC,CCT P75P NP_777557.1
XM_017027663.1 225 Silent Mutation CCC,CCT P91P XP_016883152.1

View Full Product Details