Product Details
- SNP ID
-
rs187032983
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:3433775 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGGGCATAGCACAGGTTTTGCT[A/G]CTCTGGTTGGATGGGCAGGCCCAGG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR1E2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs769431] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR1E2
- Gene Name
- olfactory receptor family 1 subfamily E member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003554.2 |
67 |
Nonsense Mutation |
CAG,TAG |
Q23* |
NP_003545.1 |
- Gene
- SPATA22
- Gene Name
- spermatogenesis associated 22
There are no transcripts associated with this gene.
View Full Product Details