Product Details

SNP ID
rs187417675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:91374958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCACGCGGATTATACTACAGTC[A/G]AGGATAGAGGGAGAAACTGGTGGAA
Phenotype
MIM: 612464
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARRDC3 PubMed Links

Gene Details

Gene
ARRDC3
Gene Name
arrestin domain containing 3
There are no transcripts associated with this gene.

Gene
ARRDC3-AS1
Gene Name
ARRDC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details