Product Details
- SNP ID
-
rs188096827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:96500850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTACAGTGCCACTTCTTTGTTT[A/G]ATATCTGGACTAAATACGCCCCCAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP54
PubMed Links
Gene Details
- Gene
- CFAP54
- Gene Name
- cilia and flagella associated protein 54
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306084.1 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
NP_001293013.1 |
XM_011539069.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537371.1 |
XM_011539070.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537372.1 |
XM_011539071.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537373.1 |
XM_011539072.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537374.1 |
XM_011539073.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537375.1 |
XM_011539074.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537376.1 |
XM_011539076.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537378.1 |
XM_011539077.2 |
378 |
Intron |
|
|
XP_011537379.1 |
XM_011539078.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537380.1 |
XM_011539079.1 |
378 |
Intron |
|
|
XP_011537381.1 |
XM_011539081.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537383.1 |
XM_011539083.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537385.1 |
XM_011539084.2 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_011537386.1 |
XM_011539085.1 |
378 |
Intron |
|
|
XP_011537387.1 |
XM_017018866.1 |
378 |
Missense Mutation |
AAT,GAT |
N112D |
XP_016874355.1 |
XM_017018867.1 |
378 |
Intron |
|
|
XP_016874356.1 |
View Full Product Details