Product Details

SNP ID
rs187863107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49628050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATCTTATTATCAGTAACTTGAA[A/G]AACTTCAATTAATGGTGGGGTCAAG
Phenotype
MIM: 612517 MIM: 602616 MIM: 180469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAAF2 PubMed Links

Gene Details

Gene
DNAAF2
Gene Name
dynein axonemal assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083908.1 2050 Intron NP_001077377.1
NM_018139.2 2050 Missense Mutation CTT,TTT L657F NP_060609.2
Gene
MGAT2
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.

Gene
RPL36AL
Gene Name
ribosomal protein L36a like
There are no transcripts associated with this gene.

View Full Product Details